siRNA Details
| Gene Symbol: Pomt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145145.1 | NP_660127.1 | 20 | 2130 | ||
| GenBank mRNA | AK053889.1 | - | 19 | 2650 | ||
| AK134770.1 | - | 20 | 2174 | |||
| AK153984.1 | - | 20 | 2229 | |||
| AY007238.1 | - | 20 | 2170 | |||
| AY494857.1 | - | 20 | 2190 | |||
| BC027325.1 | - | 20 | 2130 | 3981366 | ||

Pomt1
Genomic Map