siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290733.1 | NP_001277662.1 | 21 | 2863 | ||
| NM_001290734.1 | NP_001277663.1 | 19 | 2530 | |||
| NM_145525.3 | NP_663500.2 | 21 | 2647 | |||
| GenBank mRNA | AK037703.1 | - | 19 | 2481 | ||
| AK044411.1 | - | 21 | 2834 | |||
| AK220498.1 | - | 21 | 2594 | |||
| BC022908.1 | - | 14 | 1707 | 5043261 | ||

Osbpl6
Genomic Map