siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290733.1 | NP_001277662.1 | 14 | 1932 | ||
| NM_001290734.1 | NP_001277663.1 | 12 | 1599 | |||
| NM_145525.3 | NP_663500.2 | 13 | 1692 | |||
| GenBank mRNA | AK037703.1 | - | 12 | 1550 | ||
| AK044411.1 | - | 14 | 1903 | |||
| AK220498.1 | - | 13 | 1639 | |||
| BC022908.1 | - | 7 | 776 | 5043261 | ||

Osbpl6
Genomic Map