siRNA Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207298.2 | NP_997181.1 | 4 | 480 | ||
| GenBank mRNA | AK155384.1 | - | 4 | 476 | ||
| BB621603.1 | - | 4 | 480 | |||
| BC052176.1 | - | 4 | 441 | |||
| BC138847.1 | - | 4 | 459 | 8861869 | ||

Cercam
Genomic Map