siRNA Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026992.3 | NP_081268.1 | 3 | 694 | ||
| GenBank mRNA | AK006521.1 | - | 3 | 695 | ||
| AY028460.1 | - | 3 | 376 | |||
| BC030072.1 | - | 3 | 402 | 3376358 | ||
| BC091774.1 | - | 3 | 260 | 5150845 | ||

Dnajc24
Genomic Map