siRNA Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026992.3 | NP_081268.1 | 2 | 564 | ||
| GenBank mRNA | AK006521.1 | - | 2 | 565 | ||
| AK011571.1 | - | 2 | 278 | |||
| AY028460.1 | - | 2 | 246 | |||
| BC030072.1 | - | 2 | 272 | 3376358 | ||
| BC091774.1 | - | 2 | 130 | 5150845 | ||

Dnajc24
Genomic Map