siRNA Details
| Gene Symbol: Golga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080968.1 | NP_001074437.1 | 15 | 1250 | ||
| NM_133852.2 | NP_598613.2 | 16 | 1331 | |||
| GenBank mRNA | AK076370.1 | - | 10 | 949 | ||
| AK077516.1 | - | 15 | 1252 | |||
| AK220509.1 | - | 16 | 1259 | |||
| BC011407.1 | - | 14 | 1051 | 3596079 | ||

Golga2
Genomic Map