siRNA Details
| Gene Symbol: Usp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133857.3 | NP_598618.1 | 16 | 3226 | ||
| GenBank mRNA | AK036876.1 | - | 16 | 3227 | ||
| BC022221.1 | - | 6 | 1368 | |||
| BC132339.1 | - | 15 | 2959 | 40130616 | ||
| BC145707.1 | - | 15 | 2959 | 40131010 | ||

Usp53
Genomic Map