siRNA Details
| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170778.2 | NP_740748.1 | 10 | 1201 | ||
| GenBank mRNA | BC039699.1 | - | 10 | 1202 | 4235971 | |
| BC042543.1 | - | 6 | 666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpyd| Gene Symbol: Dpyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170778.2 | NP_740748.1 | 10 | 1201 | ||
| GenBank mRNA | BC039699.1 | - | 10 | 1202 | 4235971 | |
| BC042543.1 | - | 6 | 666 | |||