siRNA Details
Gene Symbol: Kcnc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145922.2 | NP_666034.1 | 2, 3 | 1639 | ||
GenBank mRNA | AK158073.1 | - | 2, 3 | 1039 | ||
BC024837.1 | - | 2, 3 | 1639 | 5358950 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145922.2 | NP_666034.1 | 2, 3 | 1639 | ||
GenBank mRNA | AK158073.1 | - | 2, 3 | 1039 | ||
BC024837.1 | - | 2, 3 | 1639 | 5358950 |