siRNA Details
| Gene Symbol: Prmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178891.5 | NP_849222.3 | 1 | 910 | ||
| GenBank mRNA | AK034732.1 | - | 1 | 882 | ||
| AK075617.1 | - | 1 | 366 | |||
| AK087551.1 | - | 1 | 455 | |||
| AK087608.1 | - | 1 | 909 | |||
| AK157132.1 | - | 2 | 864 | |||
| AK172003.1 | - | 1 | 902 | |||
| AK172105.1 | - | 1 | 910 | |||
| AK172722.1 | - | 1 | 910 | |||
| BC022899.1 | - | 1 | 894 | 5067159 | ||
| BC066221.1 | - | 1 | 887 | 30436952 | ||

Prmt6
Genomic Map