siRNA Details
| Gene Symbol: Ifi44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133871.2 | NP_598632.2 | 5, 6 | 1001 | ||
| GenBank mRNA | AJ299405.1 | - | 4, 5 | 891 | ||
| AK079749.1 | - | 5, 6 | 953 | |||
| AK136553.1 | - | 5, 6 | 953 | |||
| AK151346.1 | - | 5, 6 | 952 | |||
| AK171676.1 | - | 5, 6 | 934 | |||
| BC111882.1 | - | 4, 5 | 891 | 40060245 | ||
| BC112402.1 | - | 4, 5 | 891 | |||

Ifi44
Genomic Map