siRNA Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178892.5 | NP_849223.2 | 4 | 1461 | ||
| GenBank mRNA | AK028529.1 | - | 4 | 1462 | ||
| AK032453.1 | - | 4 | 1445 | |||
| AK150248.1 | - | 4 | 1443 | |||
| AK162094.1 | - | 4 | 1467 | |||
| AK163347.1 | - | 4 | 1501 | |||
| BC003919.1 | - | 3 | 721 | |||
| BC068173.1 | - | 4 | 1435 | 30616032 | ||

Tiparp
Genomic Map