siRNA Details
| Gene Symbol: Camta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081557.3 | NP_001075026.1 | 11, 12 | 3013 | ||
| GenBank mRNA | AK122383.1 | - | 6, 7 | 2439 | ||
| BC150740.1 | - | 10, 11 | 2862 | 9087651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camta1| Gene Symbol: Camta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081557.3 | NP_001075026.1 | 11, 12 | 3013 | ||
| GenBank mRNA | AK122383.1 | - | 6, 7 | 2439 | ||
| BC150740.1 | - | 10, 11 | 2862 | 9087651 | ||