siRNA Details
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153565.2 | NP_705793.1 | 2 | 694 | ||
GenBank mRNA | AK043077.1 | - | 2 | 544 | ||
AK149520.1 | - | 2 | 473 | |||
AY273821.1 | - | 2 | 318 | |||
BC038085.1 | - | 2 | 709 | 4502199 |
