siRNA Details
| Gene Symbol: Phactr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175306.4 | NP_780515.2 | 3, 4 | 338 | ||
| GenBank mRNA | AK049209.1 | - | 3, 4 | 301 | ||
| BC075672.1 | - | 3, 4 | 300 | 30549496 | ||
| BC096033.1 | - | 3, 4 | 257 | 6854767 | ||

Phactr4
Genomic Map