siRNA Details
| Gene Symbol: Phactr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161797.1 | NP_001155269.1 | 12 | 2098 | ||
| NM_175306.4 | NP_780515.2 | 13 | 2179 | |||
| GenBank mRNA | AK049209.1 | - | 13 | 2142 | ||
| AK220496.1 | - | 12 | 2077 | |||
| BC037059.1 | - | 3 | 265 | |||
| BC075672.1 | - | 13 | 2141 | 30549496 | ||
| BC096033.1 | - | 13 | 2098 | 6854767 | ||

Phactr4
Genomic Map