siRNA Details
| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089194.1 | - | 746 | |||
| AK129468.1 | - | 738 | ||||
| AK158313.1 | - | 3 | 650 | |||
| AK159547.1 | - | 821 | ||||
| AK170885.1 | - | 713 | ||||
| BC092227.1 | - | 2 | 602 | 30531788 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akna| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089194.1 | - | 746 | |||
| AK129468.1 | - | 738 | ||||
| AK158313.1 | - | 3 | 650 | |||
| AK159547.1 | - | 821 | ||||
| AK170885.1 | - | 713 | ||||
| BC092227.1 | - | 2 | 602 | 30531788 | ||