siRNA Details
| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029701.1 | - | 9 | 1722 | ||
| AK154663.1 | - | 15 | 2898 | |||
| BC036324.1 | - | 7 | 1157 | |||
| BC092227.1 | - | 16 | 3543 | 30531788 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akna| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029701.1 | - | 9 | 1722 | ||
| AK154663.1 | - | 15 | 2898 | |||
| BC036324.1 | - | 7 | 1157 | |||
| BC092227.1 | - | 16 | 3543 | 30531788 | ||