siRNA Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083592.1 | NP_001077061.1 | 5 | 964 | ||
| NM_005012.3 | NP_005003.2 | 5 | 964 | |||
| XM_011541526.1 | XP_011539828.1 | 5 | 563 | |||
| XM_017001376.1 | XP_016856865.1 | 4 | 864 | |||
| XM_017001377.1 | XP_016856866.1 | 7 | 3507 | |||
| GenBank mRNA | AK308088.1 | - | 5 | 970 | ||
| AK309189.1 | - | 2 | 206 | |||
| BC080541.1 | - | 5 | 964 | |||
| BC128386.1 | - | 6 | 574 | |||
| M97675.1 | - | 5 | 940 | |||

ROR1
Genomic Map