siRNA Details
| Gene Symbol: Lao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133892.4 | NP_598653.3 | 7 | 1428 | ||
| GenBank mRNA | AB034801.2 | - | 7 | 1428 | ||
| AK052781.1 | - | 7 | 1399 | |||
| BC017599.1 | - | 7 | 1817 | 4487137 | ||
| BC023198.1 | - | 6 | 1274 | |||

Lao1
Genomic Map