siRNA Details
| Gene Symbol: Lao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB034801.2 | - | 4 | 438 | ||
| BC017599.1 | - | 4 | 827 | 4487137 | ||
| BC023198.1 | - | 3 | 284 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lao1| Gene Symbol: Lao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB034801.2 | - | 4 | 438 | ||
| BC017599.1 | - | 4 | 827 | 4487137 | ||
| BC023198.1 | - | 3 | 284 | |||