siRNA Details
| Gene Symbol: Zfp518b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081144.2 | NP_001074613.2 | 4 | 2791 | ||
| NM_001177902.1 | NP_001171373.1 | 3 | 2725 | |||
| GenBank mRNA | AK078507.1 | - | 1 | 35 | ||
| AK173245.1 | - | 1 | 1769 | |||
| BC138368.1 | - | 1 | 2150 | 8861388 | ||
| BC138369.1 | - | 1 | 2210 | 8861389 | ||

Zfp518b
Genomic Map