siRNA Details
| Gene Symbol: Slc15a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133895.1 | NP_598656.1 | 2 | 615 | ||
| GenBank mRNA | AK048529.1 | - | 2 | 639 | ||
| AK154238.1 | - | 2 | 642 | |||
| AK182965.1 | - | 2 | 108 | |||
| AK199205.1 | - | 2 | 108 | |||
| AK214917.1 | - | 2 | 108 | |||
| AY050630.1 | - | 2 | 621 | |||
| BC016233.1 | - | 2 | 615 | 4502184 | ||

Slc15a4
Genomic Map