siRNA Details
| Gene Symbol: Lrrc8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133897.2 | NP_598658.1 | 4 | 2186 | ||
| GenBank mRNA | AB081508.1 | - | 4 | 2186 | ||
| AK089024.1 | - | 4 | 2254 | |||
| AK142337.1 | - | 4 | 2216 | |||
| AK165201.1 | - | 1 | 1206 | |||
| AK171313.1 | - | 4 | 2268 | |||
| BC019809.1 | - | 1 | 403 | |||
| BC025473.1 | - | 1 | 1707 | 5353064 | ||
| BC026572.1 | - | 4 | 2196 | 4988595 | ||

Lrrc8c
Genomic Map