siRNA Details
| Gene Symbol: Lrrc8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133897.2 | NP_598658.1 | 4 | 1239 | ||
| GenBank mRNA | AB081508.1 | - | 4 | 1239 | ||
| AK089024.1 | - | 4 | 1307 | |||
| AK142337.1 | - | 4 | 1269 | |||
| AK165201.1 | - | 1 | 259 | |||
| AK169492.1 | - | 4 | 1302 | |||
| AK171313.1 | - | 4 | 1321 | |||
| BC025473.1 | - | 1 | 760 | 5353064 | ||
| BC026572.1 | - | 4 | 1249 | 4988595 | ||

Lrrc8c
Genomic Map