siRNA Details
| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133900.4 | NP_598661.1 | 5 | 795 | ||
| GenBank mRNA | AK088865.1 | - | 5 | 747 | ||
| AK168048.1 | - | 3 | 403 | |||
| AK213907.1 | - | 2 | 44 | |||
| BC002251.1 | - | 3 | 366 | 3493291 | ||
| BQ177662.1 | - | 3 | 361 | |||

Psph
Genomic Map