siRNA Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033202.3 | NP_001028374.1 | 11 | 1138 | ||
| GenBank mRNA | AK078164.1 | - | 7 | 490 | ||
| AK144578.1 | - | 11 | 1139 | |||
| AK162153.1 | - | 187 | ||||
| AK162269.1 | - | 4 | 308 | |||
| BC038606.1 | - | 11 | 1160 | |||

Usp30
Genomic Map