siRNA Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033202.3 | NP_001028374.1 | 7, 8 | 825 | ||
| GenBank mRNA | AK078164.1 | - | 3, 4 | 177 | ||
| AK144578.1 | - | 7, 8 | 826 | |||
| BC038606.1 | - | 7, 8 | 847 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp30| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033202.3 | NP_001028374.1 | 7, 8 | 825 | ||
| GenBank mRNA | AK078164.1 | - | 3, 4 | 177 | ||
| AK144578.1 | - | 7, 8 | 826 | |||
| BC038606.1 | - | 7, 8 | 847 | |||