siRNA Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133907.3 | NP_598668.1 | 7 | 967 | ||
| GenBank mRNA | AK029973.1 | - | 7 | 777 | ||
| AK045211.1 | - | 7 | 765 | |||
| AK046056.1 | - | 7 | 968 | |||
| AK122187.1 | - | 7 | 902 | |||
| BC059245.1 | - | 7 | 782 | |||
| BC120731.1 | - | 7 | 714 | 40129654 | ||
| BC137626.1 | - | 7 | 714 | 8860645 | ||

Ube3c
Genomic Map