siRNA Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133907.3 | NP_598668.1 | 17 | 2392 | ||
| GenBank mRNA | AK029973.1 | - | 17 | 2202 | ||
| AK046056.1 | - | 17 | 2393 | |||
| AK122187.1 | - | 17 | 2327 | |||
| BC021525.1 | - | 6 | 637 | |||
| BC059245.1 | - | 17 | 2207 | |||
| BC120731.1 | - | 17 | 2139 | 40129654 | ||
| BC137626.1 | - | 17 | 2139 | 8860645 | ||

Ube3c
Genomic Map