siRNA Details
| Gene Symbol: Emilin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133918.2 | NP_598679.1 | 4 | 655 | ||
| GenBank mRNA | AK020015.1 | - | 2 | 355 | ||
| AK029337.1 | - | 4 | 626 | |||
| AK052368.1 | - | 4 | 736 | |||
| AK088995.1 | - | 4 | 719 | |||
| AK138020.1 | - | 2 | 330 | |||
| AK140388.1 | - | 2 | 71 | |||
| AK155486.1 | - | 4 | 957 | |||
| BC005481.1 | - | 4 | 655 | 3485057 | ||

Emilin1
Genomic Map