siRNA Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027295.2 | NP_081571.1 | 5 | 583 | ||
| GenBank mRNA | AB232624.1 | - | 5 | 407 | ||
| AK012286.1 | - | 5 | 586 | |||
| AK159493.1 | - | 5 | 584 | |||
| AK161391.1 | - | 5 | 464 | |||
| AK215070.1 | - | 4 | 319 | |||
| BC004580.1 | - | 5 | 583 | 3587008 | ||

Rab28
Genomic Map