siRNA Details
| Gene Symbol: Rab28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027295.2 | NP_081571.1 | 3, 4 | 447 | ||
| GenBank mRNA | AB232624.1 | - | 3, 4 | 271 | ||
| AK012286.1 | - | 3, 4 | 450 | |||
| AK159493.1 | - | 3, 4 | 448 | |||
| AK161391.1 | - | 3, 4 | 328 | |||
| BC004580.1 | - | 3, 4 | 447 | 3587008 | ||

Rab28
Genomic Map