siRNA Details
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133921.2 | NP_598682.2 | 13 | 1669 | ||
GenBank mRNA | AK132320.1 | - | 12 | 1579 | ||
DV657577.1 | - | 7 | 620 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nfxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133921.2 | NP_598682.2 | 13 | 1669 | ||
GenBank mRNA | AK132320.1 | - | 12 | 1579 | ||
DV657577.1 | - | 7 | 620 |