siRNA Details
| Gene Symbol: Pot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133931.4 | NP_598692.1 | 5 | 491 | ||
| GenBank mRNA | AK005318.1 | - | 4 | 463 | ||
| AK036052.1 | - | 5 | 492 | |||
| AK040864.1 | - | 5 | 484 | |||
| AK085041.1 | - | 5 | 482 | |||
| AK135346.1 | - | 5 | 461 | |||
| AK137376.1 | - | 5 | 461 | |||
| BC016121.1 | - | 5 | 453 | 4501733 | ||

Pot1a
Genomic Map