siRNA Details
| Gene Symbol: Tra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198102.2 | NP_932770.2 | 6 | 901 | ||
| GenBank mRNA | AK164994.1 | - | 1 | 42 | ||
| AK171708.1 | - | 5 | 2121 | |||
| BC023018.1 | - | 3 | 281 | |||
| BC057448.1 | - | 6 | 862 | 4016620 | ||
| BC058764.1 | - | 7 | 1166 | |||

Tra2a
Genomic Map