siRNA Details
| Gene Symbol: Phrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081118.1 | NP_001074587.1 | 9 | 1067 | ||
| GenBank mRNA | BC138446.1 | - | 4 | 713 | 8861466 | |
| BC145719.1 | - | 4 | 713 | 40131022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phrf1| Gene Symbol: Phrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081118.1 | NP_001074587.1 | 9 | 1067 | ||
| GenBank mRNA | BC138446.1 | - | 4 | 713 | 8861466 | |
| BC145719.1 | - | 4 | 713 | 40131022 | ||