siRNA Details
| Gene Symbol: Plekha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133942.2 | NP_598703.1 | 2 | 170 | ||
| GenBank mRNA | AK044224.1 | - | 2 | 178 | ||
| AK053879.1 | - | 2 | 214 | |||
| AK083340.1 | - | 2 | 200 | |||
| BC020017.1 | - | 2 | 170 | 3589001 | ||

Plekha1
Genomic Map