siRNA Details
| Gene Symbol: Zfp575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033205.3 | NP_001028377.1 | 4 | 1224 | ||
| GenBank mRNA | AK138180.1 | - | 4 | 1183 | ||
| AK159025.1 | - | 4 | 1225 | |||
| BC150867.1 | - | 4 | 1190 | 9087778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp575| Gene Symbol: Zfp575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033205.3 | NP_001028377.1 | 4 | 1224 | ||
| GenBank mRNA | AK138180.1 | - | 4 | 1183 | ||
| AK159025.1 | - | 4 | 1225 | |||
| BC150867.1 | - | 4 | 1190 | 9087778 | ||