siRNA Details
| Gene Symbol: AI467606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178901.3 | NP_849232.1 | 2 | 272 | ||
| GenBank mRNA | AK041954.1 | - | 2 | 328 | ||
| AK052739.1 | - | 2 | 273 | |||
| AK155213.1 | - | 2 | 327 | |||
| AK168260.1 | - | 2 | 274 | |||
| AK169672.1 | - | 2 | 351 | |||
| AK170238.1 | - | 2 | 284 | |||
| BC038694.1 | - | 2 | 258 | 1513939 | ||
| BC064101.1 | - | 2 | 259 | 1366571 | ||

AI467606
Genomic Map