siRNA Details
| Gene Symbol: Trim68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089546.1 | - | 5 | 1086 | ||
| AK155267.1 | - | 5 | 1086 | |||
| AK164544.1 | - | 5 | 1031 | |||
| BC034249.1 | - | 6 | 1037 | 3675490 | ||
| CO424835.1 | - | 4 | 516 | |||
| CX220787.1 | - | 3 | 140 | |||

Trim68
Genomic Map