siRNA Details
| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242349.1 | NP_001229278.1 | 4 | 765 | ||
| NM_178642.5 | NP_848757.4 | 4 | 762 | |||
| GenBank mRNA | AK052589.1 | - | 4 | 714 | ||
| BC062959.1 | - | 4 | 651 | 30547439 | ||

Ano1
Genomic Map