siRNA Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027256.2 | NP_081532.1 | 11, 12 | 1524 | ||
| GenBank mRNA | AK163383.1 | - | 11, 12 | 1480 | ||
| BC013710.1 | - | 11, 12 | 1470 | 3156446 | ||
| BC021838.1 | - | 4, 5 | 527 | |||
| BC031145.1 | - | 5, 6 | 603 | |||
| CF724981.1 | - | 2, 3 | 311 | |||

Ints4
Genomic Map