siRNA Details
| Gene Symbol: D8Ertd738e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146163.1 | - | 3 | 195 | ||
| AK167785.1 | - | 3 | 196 | |||
| BC024662.1 | - | 3 | 186 | 4205912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D8Ertd738e| Gene Symbol: D8Ertd738e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146163.1 | - | 3 | 195 | ||
| AK167785.1 | - | 3 | 196 | |||
| BC024662.1 | - | 3 | 186 | 4205912 | ||