siRNA Details
| Gene Symbol: D8Ertd738e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007571.2 | NP_001007572.1 | 3 | 297 | ||
| GenBank mRNA | AK146163.1 | - | 3 | 255 | ||
| AK167785.1 | - | 3 | 256 | |||
| AK215593.1 | - | 1 | 57 | |||
| BC024662.1 | - | 3 | 246 | 4205912 | ||
| CA318976.1 | - | 3 | 237 | |||
| CF740495.1 | - | 3 | 297 | |||

D8Ertd738e
Genomic Map