siRNA Details
| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133954.2 | NP_598715.2 | 2 | 340 | ||
| GenBank mRNA | AK135527.1 | - | 2 | 333 | ||
| AK160901.1 | - | 2 | 323 | |||
| AK168683.1 | - | 2 | 322 | |||
| BC016418.1 | - | 2 | 284 | 4224590 | ||

Usb1
Genomic Map