siRNA Details
| Gene Symbol: Smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012667.2 | NP_001012685.2 | 3 | 539 | ||
| NM_001146117.1 | NP_001139589.1 | 2 | 480 | |||
| GenBank mRNA | AK131902.1 | - | 2 | 231 | ||
| AK133712.1 | - | 2 | 484 | |||
| AK140957.1 | - | 2 | 270 | |||
| AK213440.1 | - | 1 | 75 | |||
| AK218393.1 | - | 1 | 96 | |||
| BB618223.1 | - | 2 | 480 | |||
| BC048089.1 | - | 3 | 515 | 6466443 | ||
| BQ922100.1 | - | 3 | 513 | |||

Smim19
Genomic Map