siRNA Details
Gene Symbol: Plekhg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081333.1 | NP_001074802.1 | 13 | 2234 | ||
GenBank mRNA | AK029859.1 | - | 11 | 2906 | ||
AK131125.1 | - | 10 | 2580 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plekhg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081333.1 | NP_001074802.1 | 13 | 2234 | ||
GenBank mRNA | AK029859.1 | - | 11 | 2906 | ||
AK131125.1 | - | 10 | 2580 |