siRNA Details
| Gene Symbol: Enkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198299.1 | NP_938041.1 | 7 | 1008 | ||
| GenBank mRNA | AK087475.1 | - | 6 | 1171 | ||
| BC018168.1 | - | 6 | 1294 | |||
| BC023921.1 | - | 6 | 1397 | |||
| BC026993.1 | - | 4 | 3075 | |||
| BC052498.1 | - | 7 | 1008 | 1245862 | ||

Enkd1
Genomic Map